Microbial Count Plate
Escherichia coli O157 count plate, Blue, green colony, 36±1 °C, 36h incubation
Catalog # 78172-714
New Product
Supplier: GVS FILTER TECHNOLOGY
Supplier #: MCPG02725
Jump to:
Product Details & Documents
GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.
- Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required
GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.
Specifications
Specifications
- Description:Escherichia coli O157 count plate
- Format:Blue, green colony, 36±1 °C, 36h incubation
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



