Microbial Count Plate
Listeria count plate, Green colony, 36±1 °C, 24±2h incubation
Catalog # 78172-624
New Product
Supplier: GVS FILTER TECHNOLOGY
Supplier #: MCPG00525
Jump to:
Product Details & Documents
GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.
- Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required
GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.
Specifications
Specifications
- Description:Listeria count plate
- Format:Green colony, 36±1 °C, 24±2h incubation



