Order Entry
Canada
Orders LinkContactUsLinkComponent
Multiple Sclerosis CD19+ B Cells

Multiple Sclerosis CD19+ B Cells

Multiple Sclerosis CD19+ B cells, Cryopreserved, Single donor, 2.5×10⁷ cells (Vial), Human, adult
Catalog # 78170-812
New Product
Supplier #:  MUS-CD19-25M
Multiple Sclerosis CD19+ B Cells
Catalog # 78170-812
New Product
CAS Number:  
Multiple Sclerosis CD19+ B Cells

Multiple Sclerosis CD19+ B cells, Cryopreserved, Single donor, 2.5×10⁷ cells (Vial), Human, adult

CAS Number:  
Price
$6,076.99
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Isolated CD19+ B cells from Multiple Sclerosis donors for studies of B-cell activation and autoantibody pathways.

  • Disease-relevant CD19+ B cells
  • Cryopreserved; high viability
  • Supports activation, autoantibody, and culture assays
  • Documentation available

CD19+ B cells Multiple Sclerosis donors are processed to preserve phenotype and function. The format enables flow/mass cytometry, culture, and molecular profiling focused on B-cell dysregulation and therapeutic evaluation.

Documents

Specifications

Specifications

  • Cell Format:
    Cryopreserved
  • Single Donor/Pooled:
    Single donor
  • Description:
    Multiple Sclerosis CD19+ B cells
  • Size:
    2.5×10⁷ cells (Vial)
  • Source:
    Human, adult

Specifications