Multiple Sclerosis CD19+ B Cells
Multiple Sclerosis CD19+ B cells, Cryopreserved, Single donor, 1×10⁷ cells (Vial), Human, adult
Catalog # 78170-466
New Product
Supplier: SANGUINE BIOSCIENCES INC
Supplier #: MUS-CD19-10M
Multiple Sclerosis CD19+ B Cells
Catalog # 78170-466
Supplier: SANGUINE BIOSCIENCES INC
New Product
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Isolated CD19+ B cells from Multiple Sclerosis donors for studies of B-cell activation and autoantibody pathways.
- Disease-relevant CD19+ B cells
- Cryopreserved; high viability
- Supports activation, autoantibody, and culture assays
- Documentation available
CD19+ B cells Multiple Sclerosis donors are processed to preserve phenotype and function. The format enables flow/mass cytometry, culture, and molecular profiling focused on B-cell dysregulation and therapeutic evaluation.
Documents
Sanguine Biospecimens.pdf
Product Literature
Specifications
Specifications
- Cell Format:Cryopreserved
- Single Donor/Pooled:Single donor
- Description:Multiple Sclerosis CD19+ B cells
- Size:1×10⁷ cells (Vial)
- Source:Human, adult
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



