pH Buffer Solutions
Buffer pH 7, yellow poly bottle, 4 L, ±0.01, 25 °C, Plastic bottle, 4 L, 7, Yellow
Catalog # CA11033-644
New Product
Supplier: ACP Chemicals Inc
Supplier #: S8781-2.5L
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Color:Yellow
- Description:Buffer pH 7, yellow poly bottle, 4 L
- Size:4 L
- Reference temperature:25 °C
- pH:7
- pH accuracy:±0.01
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



