pH Buffer Solutions
Buffer pH 4, red poly bottle, 500 ml, ±0.01, Potassium biphthalate , 25 °C, Plastic bottle, 500 ml, 4, Red
Catalog # CA11033-604
New Product
Supplier: ACP Chemicals Inc
Supplier #: S5662-500G
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Color:Red
- Description:Buffer pH 4, red poly bottle, 500 ml
- Size:500 ml
- pH buffer type:Potassium biphthalate
- Reference temperature:25 °C
- pH:4
- pH accuracy:±0.01



