Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions

pH Buffer Solutions

Buffer pH 10, blue poly bottle, 500 ml, ±0.01, Borate, 25 °C, Plastic bottle, 500 ml, 10, Blue
Catalog # CA11033-592
New Product
Supplier:   ACP Chemicals Inc
Supplier #:  H6100-2.5L
pH Buffer Solutions
Catalog # CA11033-592
Supplier:  ACP Chemicals Inc
New Product
CAS Number:  
pH Buffer Solutions
pH Buffer Solutions

Buffer pH 10, blue poly bottle, 500 ml, ±0.01, Borate, 25 °C, Plastic bottle, 500 ml, 10, Blue

CAS Number:  
Price
$120.57
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Color:
    Blue
  • Description:
    Buffer pH 10, blue poly bottle, 500 ml
  • Size:
    500 ml
  • pH buffer type:
    Borate
  • Reference temperature:
    25 °C
  • pH:
    10
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.