Orders LinkContactUsLinkComponent
Vernier® Salinity Sensor
Vernire salinitysStandard 35 ppt, 500 ml
Catalog # CA470219-326
New Product
 :  SAL-ST
Vernier® Salinity Sensor
Catalog # CA470219-326
New Product
Vernier® Salinity Sensor
Vernier® Salinity Sensor
Vernire salinitysStandard 35 ppt, 500 ml
Price
$52.09
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Easily and precisely measure the total dissolved salt content in ocean or brackish water.
Response time: 90% of full-scale reading in 10 s.
Specifications
  • Description:
    Vernire salinitysStandard 35 ppt, 500 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.