Order Entry
Canada
Orders LinkContactUsLinkComponent
Aluminium Sampling Case

Aluminium Sampling Case

Aluminium sampling case, 1250 mm
Catalog # 78117-075
New Product
Supplier #:  8509A-1200
Aluminium Sampling Case
Catalog # 78117-075
New Product
Aluminium Sampling Case
Aluminium Sampling Case

Aluminium sampling case, 1250 mm

Price
425,03 $
Log in for availability
Case of 100
Product Details & Documents

The aluminium sampling case is designed for storing samplers and equipment.

  • Available in a range of sizes
  • Lightweight and robust design
  • Foam lined to protect equipment when transported
  • Available from stock
  • Fast delivery

The sampling systems range of aluminium cases are perfect for any sampler storage requirements. They are made from robust and lightweight material and include an interior foam lining, ensuring that equipment is kept safe and protected at all times. The cases are lockable and come with a carrying handle.

Specifications
  • Description:
    Aluminium sampling case
  • Interior dimensions:
    1250 mm