Accupod Solid Samplers
Accupod sampler, sterile, HDPE, 5 ml
Catalog # 78117-014
New Product
Supplier: SAMPLING SYSTEMS LTD
Supplier #: 8360T-50S
Jump to:
Product Details & Documents
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
- Suitable for sampling blend uniformity
- Next generation in sampling technology
- Designed to minimize sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
Specifications
- Volume:5 ml
- Description:Accupod sampler, sterile
- Material:HDPE