Order Entry
Canada
Orders LinkContactUsLinkComponent
Plain Microscope Slides

Plain Microscope Slides

Plain slides, grounded edges, 45°, Soda glass, 75×25 mm, 72/box, 20 boxes/carton
Catalog # 78116-526
New Product
Supplier #:  SLD145110A
Plain Microscope Slides
Catalog # 78116-526
New Product
Plain Microscope Slides
Plain Microscope Slides

Plain slides, grounded edges, 45°, Soda glass, 75×25 mm, 72/box, 20 boxes/carton

Price
$257.11
Log in for availability
Case of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Four E's USA microscope slides are made from high-quality soda glass or super white glass.

  • Pre-cleaned, ready to use
  • Grounded edges, 45° corners
  • Autoclavable

Suitable for traditional identification labeling, these plain slides are manufactured using an advanced grinding process to ensure uniform pieces and consistency in flatness and smoothness. The polished edges and corners greatly reduce the risk of scratches and infection during use and handling of the slides.

Caution: Fragile: Handle with care.

Specifications
  • Description:
    Plain slides, grounded edges, 45°
  • L×W:
    75×25 mm
  • Material:
    Soda glass
  • Packaging:
    72/box, 20 boxes/carton

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.