Orders LinkContactUsLinkComponent
Nitrite HR Test Kits
New Product
Supplier:   CHEMetrics
Nitrite HR Test Kits
Supplier:  CHEMetrics
New Product
Certificates
Nitrite HR Test Kits
Nitrite HR Test Kits
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The NED method is widely used in the quantitative analysis of nitrite and nitrate in water samples. NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, lower detection limits and measurement ranges are possible vs. chromotropic acid kits. In addition, the nitrate measurement uses only zinc which is less toxic than cadmium, making handling and disposal safer.
  • Measurement via zinc reduction only, safer handling and disposal
    Improved sensitivity and faster analysis time
    Simplified reordering, visual refills include NED, Zn, Acidifier and buffer under single catalog code
CHEMetrics Nitrate Test Kits use the N-(1-Naphthyl) ethylenediamine (NED) method to measure nitrate in potable, surface, storm, ground, sea, and wastewater. These kits offer improved sensitivity and faster analysis time (10 vs. 13 min) compared to our current chromotropic acid-based kits.
Like the legacy chromotropic acid reagent, NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, we are now able to offer lower detection limits and measurement ranges than our chromotropic acid kits provide.
Product Family Options
Comparators
Product Information
  • Chemical method
    Range
    Application
    No. of tests
    Availability
    Price
  • 0 - 0.1 ppm
    Industrial
    Log in for availability
    Log in for availability
    Each
    Each - $31.38
    $31.38
    Each
       $31.38
    Supplier Number: C-7006
    Specifications:
    New Product
    New Product
  • 0 - 2500 ppm
    Industrial
    Log in for availability
    Log in for availability
    Each
    Each - $43.69
    $43.69
    Each
       $43.69
    Supplier Number: C-7007
    Specifications:
    New Product
    New Product
Product Information
  • Chemical method
    Range
    Application
    No. of tests
    Availability
    Price
  • NED
    0 - 250 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; boiler/chiller systems
    30
    Log in for availability
    Log in for availability
    Each
    Each - $170.55
    $170.55
    Each
       $170.55
    Supplier Number: K-7030B
    Specifications:
    New Product
    New Product
  • NED
    0 - 2500 ppm
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; boiler/chiller systems
    30
    Log in for availability
    Log in for availability
    Each
    Each - $165.09
    $165.09
    Each
       $165.09
    Supplier Number: K-7030C
    Specifications:
    New Product
    New Product
Replacement disposable pipette tip
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pipette tip, pack 30
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $10.21
    $10.21
    Pack of 30
       $10.21
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)