Order Entry
Canada
Orders LinkContactUsLinkComponent
Cold-Active TEV Protease

Cold-Active TEV Protease

1000 units, Vial
Catalog # CA78632-456
New Product
Supplier #:  P8118S
Cold-Active TEV Protease
Catalog # CA78632-456
New Product
CAS Number:  
Cold-Active TEV Protease
Cold-Active TEV Protease

1000 units, Vial

CAS Number:  
Price
$241.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cold-Active TEV Protease is an engineered version of the Tobacco Etch Virus (TEV) Protease.

  • Engineered for faster and higher performance at low temperatures

  • 3X faster than standard TEV proteases
  • Improved stability and minimal-to-no autolysis
  • Ideal for cleaving affinity tags from recombinant proteins, especially when thermosensitive
  • Contains a 7X His-tag for easy removal from samples

Cold-Active TEV Protease is optimized for higher activity at lower temperatures, improved thermal stability, and reduced autolysis. Cold-Active TEV Protease recognizes the amino-acid sequence Glu-Asn-Leu-Tyr-Phe-Gln-(Gly/Ser/Ala/Met/Cys/His/Phe) and cleaves between the Gln and Gly/Ser/Ala/Met/Cys/His/Phe residues. This enzyme is often used for the removal of affinity purification tags such as maltose-binding protein (MBP-tag) or poly-histidine (His-tag) from recombinant proteins. Cold-Active TEV Protease has a 7X His-tag for easy removal from a reaction by immobilized metal affinity chromatography (e.g. Ni Resin or Ni-NTA magnetic beads).

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    1000 units

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.