Orders LinkContactUsLinkComponent
Fentanyl Test Strips, Harm Reduction
Fentanyl Test Strips, Harm Reduction
New Product
Supplier:   W.H.P.M INC
Fentanyl Test Strips, Harm Reduction
Supplier:  W.H.P.M INC
New Product
Certificates
Fentanyl Test Strips, Harm Reduction
Fentanyl Test Strips, Harm Reduction
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Developed specifically for harm reduction use, WHPM’s fentanyl test strips have both high sensitivity and high selectivity, allowing them to detect even tiny amounts of fentanyl in drug samples without cross-reacting with non-fentanyl related substances.
  • 10 ng/ml cut-off concentration
  • Quick reference guide instructions
The WHPM fentanyl test strips were developed specifically for harm reduction use. With a detection cutoff of 10 ng/ml, they are one of the most sensitive test strips on the market. They are also highly selective to fentanyl and its analogs, meaning they are less likely to cross-react with other, non-fentanyl related drugs.
Fentanyl is a powerful synthetic opioid fifty times stronger than heroin. Since about 2018 it has become the dominant opioid on the illicit market. Found in pills and powders, including non-opioid drugs and counterfeit pharmaceuticals, fentanyl is responsible for tens of thousands of deaths a year in the U.S. alone.
Product Family Options
Product Information
  • Sensitivity
    Size
    Availability
    Price
  • 78535-048
    Fentanyl test strips
    Fentanyl test strips
    10 ng/ml
    100 tests
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $154.84
    $154.84
    Box of 100
       $154.84
    Supplier Number: WFT10-STRIP
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • 78535-044
    Fentanyl test strips
    Fentanyl test strips
    10 ng/ml
    25 tests
    Log in for availability
    Log in for availability
    Box of 25
    Box of 25 - $44.24
    $44.24
    Box of 25
       $44.24
    Supplier Number: WFT-ST25
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product