Orders LinkContactUsLinkComponent
Crimping Tools
Crimping Tools
New Product
Supplier:   Aven Tools
Crimping Tools
Supplier:  Aven Tools
New Product
Certificates
4 Options Available Below
Product Details & Documents
Aven's professional grade crimping tool frame has a smooth and efficient ratcheting mechanism that creates uniform and secure crimps. Variety of dies to extend the versatility of this tool are available. Ideal for professional electricians and DIY enthusiasts. Tool and dies are made from alloy steel from durability. 
  • The tool has a smooth ratcheting mechanism that creates uniform and secure crimps
    Ferrules and terminals are crimped by squeezing the handle through the complete cycle until the handle releases automatically
    Precise ratcheting mechanism will ensure the same amount of crimping force for all crimps
    The tool has adjustment for crimping force
    Patented design allows for quick die change without any tools
    Replacement dies available for a variety of terminals
This high-precision wire ferrule crimper features a built-in ratchet that applies correct pressure to ensure a complete crimp every time and the safety release function allows you to stop a faulty crimp before the crimping cycle is complete. The dies can easily be changed by sliding them out of the frame ‚ no tools required. Aven offers a variety of replacement dies to accommodate a wide range of wire gauges. The durable rubberized grips make this tool comfortable and easy to use.
The crimping dies are easy to replace. Simply slide the die in or out of the frame by hand. The dies snap into place with no tools required.
Product Family Options
Product Information
  • Material
    For use with
    Availability
    Price
  • 78620-816
    Crimping tool for wire ferrules, AWG 22-18/16-14/12-10, A die jaw type
    Crimping tool for wire ferrules, AWG 22-18/16-14/12-10, A die jaw type
    Alloy steel
    For Wire Ferrules AWG 22-18/16-14/12-10
    Log in for availability
    Log in for availability
    Each
    Each - 124,76 $
    124,76 $
    Each
       124,76 $
    Supplier Number: 10188
    Specifications:
    New Product
    New Product
  • 78621-122
    Crimping tool for wire ferrules, AWG 26-22/24-18/22-16, A3 die jaw type
    Crimping tool for wire ferrules, AWG 26-22/24-18/22-16, A3 die jaw type
    Alloy steel
    For Wire Ferrules AWG 26-22/24-18/22-16
    Log in for availability
    Log in for availability
    Each
    Each - 124,76 $
    124,76 $
    Each
       124,76 $
    Supplier Number: 10189
    Specifications:
    New Product
    New Product
  • 78620-892
    Crimping tool for wire ferrules, 12 – 22 AWG, D die jaw type
    Crimping tool for wire ferrules, 12 – 22 AWG, D die jaw type
    Alloy steel
    For Wire ferrules 12 - 22 AWG
    Log in for availability
    Log in for availability
    Each
    Each - 123,05 $
    123,05 $
    Each
       123,05 $
    Supplier Number: 10178
    Specifications:
    New Product
    New Product
  • 78620-342
    Crimping tool for wire ferrules, 6 - 10 AWG, F die jaw type
    Crimping tool for wire ferrules, 6 - 10 AWG, F die jaw type
    Alloy steel
    For Wire Ferrules 6 - 10 AWG
    Log in for availability
    Log in for availability
    Each
    Each - 123,05 $
    123,05 $
    Each
       123,05 $
    Supplier Number: 10179
    Specifications:
    New Product
    New Product
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Crimping die A for insulated end terminals, 22-18/16-14/12-10 AWG
    Crimping die A for insulated end terminals, 22-18/16-14/12-10 AWG
    78620-694
    Alloy steel
    Log in for availability
    Each - $50.37
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Crimping die A3 for miniature insulated terminals, 26-22/24-18/22-16 AWG
    Crimping die A3 for miniature insulated terminals, 26-22/24-18/22-16 AWG
    78620-456
    Alloy steel
    Log in for availability
    Each - $50.37
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Crimping die A1 for heat shrink terminals, 22-18/16-14/12-10/8 AWG
    Crimping die A1 for heat shrink terminals, 22-18/16-14/12-10/8 AWG
    78621-452
    Alloy steel
    Log in for availability
    Each - $50.37
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Crimping die D for cord end terminals, 22/20/18/16/14/12 AWG
    Crimping die D for cord end terminals, 22/20/18/16/14/12 AWG
    78621-136
    Alloy steel
    Log in for availability
    Each - $45.16
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Crimping die F for cord end terminals, AWG: 10/8/6
    Crimping die F for cord end terminals, AWG: 10/8/6
    78620-518
    Alloy steel
    Log in for availability
    Each - $45.16
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Crimping die F1 for cord end terminals, 4 /2 AWG
    Crimping die F1 for cord end terminals, 4 /2 AWG
    78620-720
    Alloy steel
    Log in for availability
    Each - $50.37
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.