80 LED Ring Light with Touch Control
Product Details & Documents
Featuring 80 adjustable energy-efficient LEDs, this ring light provides adjustable illumination for your inspection system.
Touch sensitive buttons provide easy brightness adjustment
Quadrant settings gives operators control over which LEDs are in use
Durable metal housing
The four touch-sensitive buttons require only minimal contact to adjust the brightness settings, reducing the possibility of jostling or loosening the ring light.
The touch sensitive buttons allow users to control the brightness and LED quadrant. The four quadrants can be individually controlled to provide users with ideal illumination over the specimen being viewed.
The 80 LED Ring Light features 4 touch-sensitive buttons controlling power, brightness, and LED quadrant illumination. The included ring light adapter screws to the bottom of all Aven stereo zoom microscopes, providing easy and secure ring light attachment.
Color temperature: 6500 to 7000 K
Brightness adjustment: Via touch sensitive buttons
Working distance: 50 to 180 mm
Delivery information: Includes 100 to 240 V power supply and a ring light adapter.
Specifications
- Description:80 LED ring light with touch control, Int. Ø: 25 - 63 mm (0.98 - 2.5")
- Electrical:100 - 240 V, 50/60 Hz, 0.2 A max
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



