Technik™ Cutter
About this item
End cutters are used to cut close to the surface of your material, leaving a smooth surface behind.
Manufactured from 410 stainless steel
Box Joint construction provides superior jaw alignment and durability
Induction hardened jaws
Dual leaf springs
ESD Safe cushioned grips
3 Options Available Below
Product Details & Documents
End cutters are used to cut close to the surface of your material, leaving a smooth surface behind.
Manufactured from 410 stainless steel
Box Joint construction provides superior jaw alignment and durability
Induction hardened jaws
Dual leaf springs
ESD Safe cushioned grips
Specifications for Product Family
Specifications
- Catalog No:
- 78620-528
- 78620-596
- 78621-052
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
Specifications
Product Family Options
Product Information
- MaterialCutting capacityBlade lengthOverall lengthJaw widthJaw typeAvailabilityPrice
- Stainless Steel 4100.2 - 1.25mm soft copper wire17.5 mm114 mm11.85 mmEnd cutterLog in for availabilityEach$39.31Supplier Number: 10327Specifications:New Product
More
Specifications
Cat. No.78620-528Disposable / reusableReusableNew Product - Stainless Steel 4100.2 - 1.25mm soft copper wire—114 mm11.85 mmOblique cutterLog in for availabilityEach$39.31Supplier Number: 10328Specifications:New Product
More
Specifications
Cat. No.78620-596Disposable / reusableReusableNew Product - Stainless Steel 4100.1 - 1.25mm soft copper wire10.3 mm110 mm9.5 mmTapered miniature cutterLog in for availabilityEach$39.31Supplier Number: 10331Specifications:New Product
More
Specifications
Cat. No.78621-052Disposable / reusableReusableNew Product