Order Entry
Canada
Orders LinkContactUsLinkComponent
Focus Mount

Focus Mount

Focus mount, 50 mm vertical direction focusing distance
Catalog # 78620-948
New Product
Supplier:   Aven Tools
Supplier #:  26800B-517
Focus Mount
Catalog # 78620-948
Supplier:  Aven Tools
New Product
Focus Mount

Focus mount, 50 mm vertical direction focusing distance

Price
$1,215.90
Log in for availability
Each
Product Details & Documents

This Focus Mount with 74 mm diameter ring will accept SSZ, SPZ, DSZ and NSW series microscopes.

  • Coarse and fine focus adjustments

    26.9 mm end hole adapter

  • Precision rack and Pinion mechanism with tension control

A reducer ring for mounting on post size of diameter 25 mm is included with the package, and a focus mount has a precision rack and pinion mechanism with tension control and coarse and fine focus knobs. A tension control adjustment pin is included with the mount.

Specifications
  • Description:
    Focus mount, 50 mm vertical direction focusing distance