Order Entry
Canada
Orders LinkContactUsLinkComponent
Formalin Titration Kit

Formalin Titration Kit

Formalin titration kit, Test kit
Catalog # CA78611-994
New Product
Supplier:   CHAPTEC INC CA
Supplier #:  TUY0001
Formalin Titration Kit
Catalog # CA78611-994
Supplier:  CHAPTEC INC CA
New Product
CAS Number:  
Formalin Titration Kit
Formalin Titration Kit

Formalin titration kit, Test kit

CAS Number:  
Price
$194.21
Log in for availability
Kit of 1 (1/Itm/Bx)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Drop-count titration kit are a simple, easy, and cost-effective way to test any formalin and TissuFix (formaldehyde in aqueous solution 3.7 to 7.4%), in the range of 10 to 20% formalin. Drop counting titration is a simple and quick way to cost-effectively test different parameters.

CAS No.: 64-17-5 (60.0 - 80.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 77-09-8 (0.1 - 1.0%); 7664-93-9 (7.0 - 13.0%); 7757-83-7 (80.0 - 100.0%)

Specifications

Specifications

  • Description:
    Formalin titration kit
  • Application:
    Neutralizer
  • Details:
    Test kit

Specifications