Order Entry
Canada
Orders LinkContactUsLinkComponent
PBS Buffer Concentrate

PBS Buffer Concentrate

PBS buffer, 132 ml, Plastic bottle, Case of 20 bottles, 7.2
Catalog # CA78612-172
New Product
Supplier:   CHAPTEC INC CA
Supplier #:  TPC3934
PBS Buffer Concentrate
Catalog # CA78612-172
Supplier:  CHAPTEC INC CA
New Product
CAS Number:  
PBS Buffer Concentrate
PBS Buffer Concentrate

PBS buffer, 132 ml, Plastic bottle, Case of 20 bottles, 7.2

CAS Number:  
Price
$197.31
Log in for availability
Case of 20 (132ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Designed for immunohistochemistry techniques, the PBS buffer is used to dilute antibodies and wash the slides.

  • Available in concentrated solution, in containers of 132 ml for 4 L of preparation
  • pH of 7.2 ±0.2
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Description:
    PBS buffer
  • Size:
    Case of 20 bottles
  • pH:
    7.2
  • Application:
    Neutralizer
  • Volume:
    132 ml
  • Cat. No.:
    CA78612-172
  • CAS No.:
    7647-14-5 (10.0 - 30.0%); 7558-79-4 (1.0 - 5.0%); 7778-77-0 (0.1 - 1.0%); 7447-40-7 (0.1 - 1.0%)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.