Order Entry
Canada
Orders LinkContactUsLinkComponent
Deolorizing Solutions 0.25%

Deolorizing Solutions 0.25%

New Product
Supplier:   CHAPTEC INC CA
Deolorizing Solutions 0.25%
Supplier:  CHAPTEC INC CA
New Product
Certificates
Deolorizing Solutions 0.25%

3 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Decolorizing solution for H&E & PAP stain.

  • Available in various concentrations of hydrochloric acid (0.25%, 0.5%, and 1%) in 70% alcohol

The 0.25% alcohol-acid differentiation solution is a 70% alcoholic solution containing 0.25% concentrated hydrochloric acid. It is commonly used in histology and cytology to remove excess stain and highlight nuclei after hematoxylin staining in the H&E and PAP methods. To achieve H&E staining with precise nuclear details, it is crucial that certain hematoxylins are properly differentiated, making the alcohol-acid solution particularly effective.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CA78612-088
  • CA78612-150
  • CA78612-168
  • CAS No.:
  • 64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)
  • 64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)
  • 64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)

Specifications

Product Family Options

Product Information

  • Application
    Volume
    Pack type
    Availability
    Price
  • CA78612-088
    Decolorizing solution 0.25%
    Decolorizing solution 0.25%
    Destaining reagents
    4 L
    Plastic jug
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $123.10
    $123.10
    Each(1/Itm/Bx)
       $123.10

    Supplier Number: DEY0025
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    CA78612-088
    CAS No.
    64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)
    New Product
  • CA78612-150
    Decolorizing solution 0.25%
    Decolorizing solution 0.25%
    Destaining reagents
    1 L
    Plastic bottle
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $25.82
    $25.82
    Each(1/Itm/Bx)
       $25.82

    Supplier Number: DEY0025-1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    CA78612-150
    CAS No.
    64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)
    New Product
  • CA78612-168
    Decolorizing solution 0.25%
    Decolorizing solution 0.25%
    Destaining reagents
    500 ml
    Plastic bottle
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $23.83
    $23.83
    Each(1/Itm/Bx)
       $23.83

    Supplier Number: DEY0025-500
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    CA78612-168
    CAS No.
    64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.