Epic Basic Protection Cleanroom Shirt
Product Details & Documents
Latex free/low lint pullover shirt for use in controlled environments and many general application areas. Breathable cloth-like fabric is comfortable and cool. Features long sleeves, elastic wrists and a vertical slit back for added comfort and user fit.
Cleanroom-compatible design: Manufactured for controlled environments where particle and contamination control is critical
- Basic protection level: Provides essential barrier against non-hazardous particulates and light liquid splashes
- Durable fabric: Made from high-quality, low-linting material to minimize contamination risk
- Comfortable fit: Ergonomic design for ease of movement during extended wear
- Secure closure: Snap or zipper front for quick donning and doffing
- Elastic cuffs: Helps maintain sleeve position and reduces particle escape
- Reusable or disposable options: Depending on product specification, supports cost-effective cleanroom operations
Essential cleanroom apparel for controlled environments requiring basic contamination control. The Tians Epic Basic Protection Cleanroom Shirt is designed to provide reliable particle and contamination barrier performance for non-critical cleanroom operations. Manufactured from low-linting, durable fabric, this shirt helps maintain cleanroom integrity while ensuring operator comfort during extended wear. Ideal for ISO Class cleanrooms and controlled environments in pharmaceutical, electronics, and medical device manufacturing, it offers a cost-effective solution for routine processes.
Garment is constructed from Environstar-SPP spunbonded polypropylene fabric which is cloth-like and has exceptional breathability. Individually inspected for sizing, stitching, and workmanship.
Benefits
Contamination control: Reduces particle shedding and helps maintain cleanroom integrity.
Operator comfort: Lightweight and breathable material enhances comfort during long shifts.
Compliance support: Meets cleanroom apparel standards for ISO Class environments.
Cost efficiency: Basic protection level ideal for non-critical processes, reducing unnecessary expense.
Versatile use: Suitable for pharmaceutical, electronics, and medical device cleanrooms.
Ball Burst: 82.58 N
Particle shedding (Helmke Drum): Level II
Water resistance: No resistance
Water vapor transmission rate: 6856 g/m²/24 hrs
Tearing strength: 3.5 lbs./in (Machine direction) 7.1 lbs./in (Cross direction)
Tensile strength: 16 lbs./in (Machine direction)
11 lbs./in (Cross direction)
Certifications: ISO 7/Class 10000.
Packaging: Neatly folded and clean room packed: 10 pieces per inner bag, 5 inner bags per master bag, 50 pieces per case.
Specifications
- Clothing Size:Universal
- Color:White
- L×W:32×29"
- Type:Shirt
- Thickness:0.21 mm
- Cat. No.:78150-658
- Sleeve or leg length:Long
- Garment application:Cleanroom
- Gender:Unisex
- Fabric:Spunbound polypropylene
- Weight:25 g/m²
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



