Orders LinkContactUsLinkComponent
Transferpette® pro, Ejector Set (upper and lower)
New Product
 :   Brandtech
Transferpette® pro, Ejector Set (upper and lower)
 :  Brandtech
New Product
Transferpette® pro, Ejector Set (upper and lower)
Transferpette® pro, Ejector Set (upper and lower)
Replacement ejector set.
  • Ejector set (upper and lower part) for use with Transferpette® pro single channel pipettors, 2.5 to 10ml
Designed for a perfect fit, simple installation, and long-lasting reliability.
Product Family Options
  • Volume
  • 1 - 10 ml
    Each
    Each - 76,55 $
    76,55 $
    Each
       76,55 $
     : 704757
    New Product
    New Product
  • 0.25 - 2.5 mL
    Each
    Each - 76,55 $
    76,55 $
    Each
       76,55 $
     : 704755
    New Product
    New Product
  • 0.5 - 5 mL
    Each
    Each - 76,55 $
    76,55 $
    Each
       76,55 $
     : 704756