Order Entry
Canada
Orders LinkContactUsLinkComponent
Drip Tray

Drip Tray

Drip tray for vial rack system, 184.9 mm, 40.1 mm
Catalog # 78473-706
New Product
Supplier:   SAVILLEX
Supplier #:  550-740-600671
Drip Tray
Catalog # 78473-706
Supplier:  SAVILLEX
New Product
Drip Tray

Drip tray for vial rack system, 184.9 mm, 40.1 mm

Price
$105.07
Log in for availability
Each
Product Details & Documents

Drip tray, for vial rack system, allows vial rack system to cool and dry without dripping acid on lab surfaces.

  • Allows vial rack system to cool and dry without dripping acid on lab surfaces

    Heavy duty construction, easy to grasp

Specifications
  • Ø:
    184.9 mm
  • Description:
    Drip tray for vial rack system
  • Height:
    40.1 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.