Column Component Closures
New Product
Supplier: SAVILLEX
Jump to:
About this item
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
9 Options Available Below
Product Details & Documents
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Product Family Options
Product Information
- AvailabilityPrice
- 78473-816Column component closure, with one ¹/₄" FNPT port, 58 mmSupplier Number: 600-058-17Specifications:New ProductNew Product
- 78474-141Column component closure, with one ³/₈" FNPT port, 58 mmSupplier Number: 600-058-19Specifications:New ProductNew Product
- 78473-538Column component closure, with two ¹/₈" outer Ø tube port, 58 mmColumn component closure, with two ¹/₈" outer Ø tube port, 58 mmLog in for availabilityEach$136.91Supplier Number: 600-058-24Specifications:New ProductNew Product
- 78473-600Column component closure, with two ¹/₄" outer Ø tube port, 58 mmColumn component closure, with two ¹/₄" outer Ø tube port, 58 mmLog in for availabilityEach$155.90Supplier Number: 600-058-33Specifications:New ProductNew Product
- 78473-842Column component closure, with two ¹/₈" push in transfer side ports, 24 mmColumn component closure, with two ¹/₈" push in transfer side ports, 24 mmLog in for availabilityEach$56.90Supplier Number: 600-024-23Specifications:New ProductNew Product
- 78474-153Column component closure, 58 mmSupplier Number: 600-058-06Specifications:New ProductNew Product
- 78473-608Column component closure, with one ¹/₄" outer Ø tube port, 58 mmSupplier Number: 600-058-16Specifications:New ProductNew Product
- 78474-155Column component closure, with two ¹/₂" outer Ø tube port, 58 mmColumn component closure, with two ¹/₂" outer Ø tube port, 58 mmLog in for availabilityEach$226.32Supplier Number: 600-058-42Specifications:New ProductNew Product
- 78474-575Transfer closure, pressure relief valve, Ultem® clamp, 1¹/₂" FNPT threads, 58 mmTransfer closure, pressure relief valve, Ultem® clamp, 1¹/₂" FNPT threads, 58 mmLog in for availabilityEach$418.79Supplier Number: 600-058-87Specifications:New ProductNew Product