Orders LinkContactUsLinkComponent
Vent Nut Assembly
Vent Nut Assembly
¼" Vent nut assembly, PTFE
Catalog # 78473-744
New Product
Supplier:   SAVILLEX
Supplier #:  550-740-300731
Vent Nut Assembly
Catalog # 78473-744
Supplier:  SAVILLEX
New Product
Vent Nut Assembly
¼" Vent nut assembly, PTFE
Price
$33.67
Log in for availability
Each
Product Details & Documents
Used to vent the vial cleaning system during heating. The adjustable vent nut assembly also enables smooth controlled liquid flow through the pour spout. Recommended annual replacement for vial cleaning system.
  • ¹/₄" vent nut
    9 mm PTFE filter membrane
    PTFE retaining ring
Delivery information: Two years warranty.
Specifications
  • Description:
    ¼" Vent nut assembly
  • Material:
    PTFE