edge® pH/ORP Meter
Product Details & Documents
Sleek, versatile pH and ORP meter designed for laboratory, classroom, or field use.
- Measures pH and oxidation-reduction potential (ORP) with professional accuracy
- Large high-contrast LCD with wide viewing angle for clear data display
- Touch-sensitive keypad and intuitive menu navigation
- Smart electrode recognition automatically loads sensor and calibration data
- Supports benchtop, wall-mount, or portable operation
- Lightweight, thin design maximizes workspace and mobility
- USB connectivity for data export and record keeping
The edge® pH/ORP Meter combines precision and modern design in a single, easy-to-use instrument that adapts to laboratory, classroom, or field applications. Its slim, lightweight profile and modular stand allow it to function as a benchtop, wall-mounted, or portable meter, providing maximum flexibility for any workspace. This advanced digital meter measures both pH and oxidation-reduction potential (ORP) using smart electrodes that automatically transmit sensor type, calibration data, and serial number to the instrument. Automatic temperature compensation ensures reliable readings across a range of sample types and conditions.
The unit features a large, backlit LCD display with a 150° viewing angle and intuitive touch-sensitive controls for effortless navigation. Automatic calibration guides users through up to three points using standard or custom buffers, while built-in electrode diagnostics help identify issues such as contamination or aging. Designed with educators and researchers in mind, the edge® meter provides laboratory-grade accuracy in a user-friendly format that suits chemistry, biology, and environmental science instruction. With a rechargeable battery, USB data transfer, and multiple mounting options, it delivers portability and performance ideal for both teaching and professional applications.
Delivery information: Rechargeable battery and 5 V DC adapter (included).
Specifications
- Description:edge® pH/ORP meter
- Cat. No.:CA470360-144
- pH accuracy:±0.01 pH
- Dimensions:202×140×12.7 mm (8×5.5×0.5")
- Weight:250 g (8.8 oz.)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



