Masterflex® L/S® Variable Speed Analog Console Pump Systems
About this item
Don’t lose control of your metering applications - digital display gives you precise control.
- ABS plastic and aluminum housing
-
Accurate and repeatable settings with three-digit LED display of rpm
- Pump fluid in either direction with reversible motor - perfect for purging tubing
- Choose multichannel systems for synchronous flow from all channels
- Maximize processes in a minimal footprint by stacking splash-resistant, IP 33-rated drives
- Two ways to control speed, direction, and start/stop - use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Certifications: CE Compliant.
45 Options Available Below
Product Details & Documents
Don’t lose control of your metering applications - digital display gives you precise control.
- ABS plastic and aluminum housing
-
Accurate and repeatable settings with three-digit LED display of rpm
- Pump fluid in either direction with reversible motor - perfect for purging tubing
- Choose multichannel systems for synchronous flow from all channels
- Maximize processes in a minimal footprint by stacking splash-resistant, IP 33-rated drives
- Two ways to control speed, direction, and start/stop - use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Certifications: CE Compliant.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- MFLX77926-20-IT
- MFLX77917-00-IT
- MFLX77917-10-IT
- MFLX77917-20-IT
- MFLX77917-30-IT
- MFLX77926-20-IL
- MFLX77917-00-IL
- MFLX77917-10-IL
- MFLX77917-20-IL
- MFLX77917-30-IL
- MFLX77917-00-CH
- MFLX77917-10-CH
- MFLX77917-20-CH
- MFLX77917-30-CH
- MFLX77926-20-CH
- MFLX77926-20-AU
- MFLX77917-00-AU
- MFLX77917-10-AU
- MFLX77917-20-AU
- MFLX77917-30-AU
- MFLX77926-20-DK
- MFLX77917-00-DK
- MFLX77917-10-DK
- MFLX77917-20-DK
- MFLX77917-30-DK
- MFLX77926-20-IN
- MFLX77917-00-IN
- MFLX77917-10-IN
- MFLX77917-20-IN
- MFLX77917-30-IN
- MFLX77926-20-EU
- MFLX77917-00-EU
- MFLX77917-10-EU
- MFLX77917-20-EU
- MFLX77917-30-EU
- MFLX77926-20-UK
- MFLX77917-00-UK
- MFLX77917-10-UK
- MFLX77917-20-UK
- MFLX77917-30-UK
- MFLX77926-20
- MFLX77917-00
- MFLX77917-10
- MFLX77917-20
- MFLX77917-30
- Model:
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77926-20
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- 77926-20
- 77917-00
- 77917-10
- 77917-20
- 77917-30
- No. of channels:
- 4
- 1
- 1
- 1
- 1
- 4
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 4
- 4
- 1
- 1
- 1
- 1
- 4
- 1
- 1
- 1
- 1
- 4
- 1
- 1
- 1
- 1
- 4
- 1
- 1
- 1
- 1
- 4
- 1
- 1
- 1
- 1
- 4
- 1
- 1
- 1
- 1
- No. of rollers:
- 6
- 3
- 3
- 3
- 3
- 6
- 3
- 3
- 3
- 3
- 3
- 3
- 3
- 3
- 6
- 6
- 3
- 3
- 3
- 3
- 6
- 3
- 3
- 3
- 3
- 6
- 3
- 3
- 3
- 3
- 6
- 3
- 3
- 3
- 3
- 6
- 3
- 3
- 3
- 3
- 6
- 3
- 3
- 3
- 3
- Control type:
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Housing material:
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- ABS plastic and aluminium
- Min. flow rate per channel:
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 4.8 ml/min
- 17 ml/min
- 4.8 ml/min
- 17 ml/min
- Max. flow rate per channel:
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 47 ml/min
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- 47 ml/min
- 480 ml/min
- 1700 ml/min
- 480 ml/min
- 1700 ml/min
- Min. flow rate:
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- 0.47 ml/min
- —
- —
- 4.8 ml/min
- 17 ml/min
- Drive IP rating:
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- Accuracy:
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
- Anti-drip:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Cloud connected:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Digital dispensing:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Drive:
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
- Duty cycle:
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Min. RPM:
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 1 rpm
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 1 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- Open-head sensor:
- No
- No
- No
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- No
- No
- Yes
- Yes
- No
- No
- No
- No
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- Power:
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Remote I/O:
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
- Reversible motor:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Tubing:
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16 tube set, pk of 8
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Masterflex® L/S® 16, 10'
- Masterflex® L/S® 24, 10'
- Tubing sizes accepted:
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® two-stop tube sets: L/S® 13, 14, 16
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- Washdown capable:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Current:
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- 1.8 A @ 115 V; 1.2 A @ 230 V
- Frequency:
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- Voltage:
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
Specifications
Product Family Options
Product Information
- Plug typeMaximum Flow RateHeightLengthWidthPump HeadMax. RPMAvailabilityPrice
- MFLX77926-20-ITL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACIT188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-ITModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-ITL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACIT480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-ITModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-ITL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACIT1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-ITModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-ITL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACIT480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-ITModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-ITL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACIT1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-ITModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20-ILL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACIL188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-ILModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-ILL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACIL480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-ILModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-ILL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACIL1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-ILModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-ILL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACIL480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-ILModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-ILL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACIL1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-ILModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-CHL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACCH480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-CHModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-CHL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACCH1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-CHModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-CHL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACCH480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-CHModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-CHL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACCH1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-CHModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20-CHL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACCH188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-CHModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20-AUL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACAU188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-AUModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-AUL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACAU480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-AUModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-AUL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACAU1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-AUModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-AUL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACAU480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-AUModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-AUL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACAU1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-AUModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20-DKL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACDK188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-DKModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-DKL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACDK480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-DKModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-DKL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACDK1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-DKModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-DKL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACDK480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-DKModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-DKL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACDK1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-DKModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20-INL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACIN188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-INModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-INL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACIN480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-INModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-INL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACIN1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-INModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-INL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACIN480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-INModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-INL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACIN1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-INModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20-EUL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACEU188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-EUModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-EUL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACEU480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-EUModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-EUL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACEU1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-EUModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-EUL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACEU480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-EUModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-EUL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACEU1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-EUModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20-UKL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACUK188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20-UKModel77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00-UKL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACUK480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00-UKModel77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10-UKL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACUK1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10-UKModel77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20-UKL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACUK480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20-UKModel77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30-UKL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACUK1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30-UKModel77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77926-20L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACL/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VACUS188 ml/min215.9 mm (8.5")349 mm (13.74")210.8 mm (8.3")07535-04 (L/S® 4-channel pump head)100 rpmLog in for availabilityEach$5,510.00Specifications:New Product
More
Specifications
Cat. No.MFLX77926-20Model77926-20No. of channels4No. of rollers6Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel0.47 ml/minMax. flow rate per channel47 ml/minMin. flow rate0.47 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)Duty cycleContinuousMin. RPM1 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16 tube set, pk of 8Tubing sizes acceptedL/S® two-stop tube sets: L/S® 13, 14, 16Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-00L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VACUS480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-60 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-00Model77917-00No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-10L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VACUS1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-62 (L/S® Easy-Load® II pump head)600 rpmLog in for availabilityEach$4,440.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-10Model77917-10No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate—Drive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-20L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VACUS480 ml/min215.9 mm (8.5")352.6 mm (13.88")210.8 mm (8.3")77200-30 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-20Model77917-20No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 16, 10'Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product - MFLX77917-30L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACL/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VACUS1700 ml/min215.9 mm (8.5")339.9 mm (13.38")210.8 mm (8.3")77200-32 (L/S® Easy-Load® II OHS pump head)600 rpmLog in for availabilityEach$4,650.00Specifications:New Product
More
Specifications
Cat. No.MFLX77917-30Model77917-30No. of channels1No. of rollers3Control typeVariable-Speed DigitalHousing materialABS plastic and aluminiumMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)Anti-dripNoCloud connectedNoDigital dispensingNoDrive07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorYesPower90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 VPump seriesL/SRemote I/O4 - 20 mA, 0 - 10 V, start/stop and direction via contact closureReversible motorYesTubingMasterflex® L/S® 24, 10'Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent1.8 A @ 115 V; 1.2 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 yearsNew Product