Multiple Sclerosis CD19+ B Cells
New Product
Isolated CD19+ B cells from Multiple Sclerosis donors for studies of B-cell activation and autoantibody pathways.
- Disease-relevant CD19+ B cells
- Cryopreserved; high viability
- Supports activation, autoantibody, and culture assays
- Documentation available
CD19+ B cells Multiple Sclerosis donors are processed to preserve phenotype and function. The format enables flow/mass cytometry, culture, and molecular profiling focused on B-cell dysregulation and therapeutic evaluation.
Isolated CD19+ B cells from Multiple Sclerosis donors for studies of B-cell activation and autoantibody pathways.
- Disease-relevant CD19+ B cells
- Cryopreserved; high viability
- Supports activation, autoantibody, and culture assays
- Documentation available
CD19+ B cells Multiple Sclerosis donors are processed to preserve phenotype and function. The format enables flow/mass cytometry, culture, and molecular profiling focused on B-cell dysregulation and therapeutic evaluation.
Product Family Options
- DescriptionCell FormatSingle Donor/PooledSizeSource
- Multiple Sclerosis CD19+ B cellsCryopreservedSingle donor1×10⁷ cells (Vial)Human, adultEach$3,376.11: MUS-CD19-10MNew ProductNew Product
- Multiple Sclerosis CD19+ B cellsCryopreservedSingle donor2.5×10⁷ cells (Vial)Human, adultEach$6,076.99: MUS-CD19-25MNew ProductNew Product