Ulcerative Colitis Serum
Ulcerative Colitis serum, human adult, single donor, 1 ml, Cryopreserved
Catalog # 78170-660
New Product
Supplier: SANGUINE BIOSCIENCES INC
Supplier #: ULC-SER-1ML
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Serum from clinically confirmed Ulcerative Colitis donors, frozen to preserve analyte integrity for biomarker and serology assays.
- Clinically confirmed Ulcerative Colitis donors
- 1 ml per vial
- Frozen; stable analytes
- Supports biomarker and serology workflows
- Documentation available
Ulcerative Colitis serum 1 ml supports antibody testing, biomarker discovery, and method validation. Standardized collection and processing help maintain consistency across longitudinal and cross-sectional studies.
Documents
Sanguine Biospecimens.pdf
Product Literature
Specifications
- Serum format:Cryopreserved
- Description:Ulcerative Colitis serum, human adult, single donor
- Size:1 ml
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.