Order Entry
Canada
Orders LinkContactUsLinkComponent
Hepatitis B Plasma

Hepatitis B Plasma

Hepatitis B plasma, single donor, 1 ml, Cryopreserved, Human adult
Catalog # 78170-810
New Product
Supplier #:  HPB-PLA-1ML
Hepatitis B Plasma
Catalog # 78170-810
New Product
CAS Number:  
Hepatitis B Plasma

Hepatitis B plasma, single donor, 1 ml, Cryopreserved, Human adult

CAS Number:  
Price
$759.63
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Plasma from clinically confirmed Hepatitis B donors, frozen for proteomic and cytokine analyses.

  • Clinically confirmed Hepatitis B donors

    1 ml per vial

    Frozen; QC-managed handling

    Suitable for proteomics and cytokine profiling

    Documentation available

Hepatitis B plasma 1 ml is processed to preserve analyte integrity for downstream assays including cytokine/chemokine panels and proteomics. Matched materials may be available for integrated study designs.

Documents

Specifications

Specifications

  • Description:
    Hepatitis B plasma, single donor
  • Size:
    1 ml
  • Format:
    Cryopreserved
  • Origin:
    Human adult

Specifications