Ulcerative Colitis Plasma
Ulcerative colitis plasma, single donor, 1 ml, Cryopreserved, Human adult
Catalog # 78170-782
New Product
Supplier: SANGUINE BIOSCIENCES INC
Supplier #: ULC-PLA-1ML
Ulcerative Colitis Plasma
Catalog # 78170-782
Supplier: SANGUINE BIOSCIENCES INC
New Product
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Plasma from clinically confirmed ulcerative colitis donors, frozen for proteomic and cytokine analyses.
Clinically confirmed ulcerative colitis donors
1 ml per vial
Frozen; QC-managed handling
Suitable for proteomics and cytokine profiling
Documentation available
Ulcerative colitis plasma 1 ml is processed to preserve analyte integrity for downstream assays including cytokine/chemokine panels and proteomics. Matched materials may be available for integrated study designs.
Documents
Sanguine Biospecimens.pdf
Product Literature
Specifications
Specifications
- Description:Ulcerative colitis plasma, single donor
- Size:1 ml
- Format:Cryopreserved
- Origin:Human adult
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



