Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Scleroderma Plasma

Scleroderma plasma, single donor, 1 ml, Cryopreserved, Human adult
Catalog # 78170-848
New Product
 :  SYS-PLA-1ML
Scleroderma Plasma
Catalog # 78170-848
New Product
CAS Number:  
Scleroderma Plasma

Scleroderma plasma, single donor, 1 ml, Cryopreserved, Human adult

CAS Number:  
Price
$759.63
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Plasma from clinically confirmed scleroderma donors, frozen for proteomic and cytokine analyses.

  • Clinically confirmed scleroderma donors

    1 ml per vial

    Frozen; QC-managed handling

    Suitable for proteomics and cytokine profiling

    Documentation available

Scleroderma plasma 1 ml is processed to preserve analyte integrity for downstream assays including cytokine/chemokine panels and proteomics. Matched materials may be available for integrated study designs.

Specifications

Specifications

  • Description:
    Scleroderma plasma, single donor
  • Size:
    1 ml
  • Format:
    Cryopreserved
  • Origin:
    Human adult

Specifications