Hypercholesterolemia Plasma
Hypercholesterolemia plasma, single donor, 1 ml, Cryopreserved, Human adult
Catalog # 78170-544
New Product
Supplier: SANGUINE BIOSCIENCES INC
Supplier #: HYC-PLA-1ML
Hypercholesterolemia Plasma
Catalog # 78170-544
Supplier: SANGUINE BIOSCIENCES INC
New Product
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Plasma from clinically confirmed high cholesterol donors, frozen for proteomic and cytokine analyses.
Clinically confirmed high cholesterol donors
1 ml per vial
Frozen; QC-managed handling
Suitable for proteomics and cytokine profiling
Documentation available
High cholesterol plasma 1 ml is processed to preserve analyte integrity for downstream assays including cytokine/chemokine panels and proteomics. Matched materials may be available for integrated study designs.
Documents
Sanguine Biospecimens.pdf
Product Literature
Specifications
Specifications
- Description:Hypercholesterolemia plasma, single donor
- Size:1 ml
- Format:Cryopreserved
- Origin:Human adult



