COPD PBMC
About this item
Cryopreserved peripheral blood mononuclear cells (PBMCs) from clinically confirmed COPD donors for immunology and translational research.
Clinically confirmed COPD donors
High-viability cryopreservation
Research-ready aliquots
Supports flow, cytokine, and gene-expression assays
Documentation available
COPD PBMC are collected under IRB oversight and processed to preserve cellular integrity and functional responses. The PBMC format is suitable for phenotyping, cytokine release assays, co-culture systems, and multi-omics workflows investigating immune mechanisms in copd.
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Cryopreserved peripheral blood mononuclear cells (PBMCs) from clinically confirmed COPD donors for immunology and translational research.
Clinically confirmed COPD donors
High-viability cryopreservation
Research-ready aliquots
Supports flow, cytokine, and gene-expression assays
Documentation available
COPD PBMC are collected under IRB oversight and processed to preserve cellular integrity and functional responses. The PBMC format is suitable for phenotyping, cytokine release assays, co-culture systems, and multi-omics workflows investigating immune mechanisms in copd.
Product Family Options
Product Information
- Cell FormatSingle Donor/PooledSizeSourceAvailabilityPrice
- 78170-682COPD PBMCCOPD PBMCCryopreservedSingle donor2×10⁹ cells (Vial, bulk)Human, adultLog in for availabilityEach$21,100.65Supplier Number: CPD-LKLT-2BItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- 78170-722COPD PBMCCOPD PBMCCryopreservedSingle donor1×10⁸ cells (Vial)Human, adultLog in for availabilityEach$1,856.86Supplier Number: CPD-LKLT-100MItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- 78170-464COPD PBMCCOPD PBMCCryopreservedSingle donor1×10⁷ cells (Vial)Human, adultLog in for availabilityEach$835.59Supplier Number: CPD-LKLT-10MItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- 78170-718COPD PBMCCOPD PBMCCryopreservedSingle donor2.5×10⁷ cells (Vial)Human, adultLog in for availabilityEach$1,181.65Supplier Number: CPD-LKLT-25MItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- 78170-558COPD PBMCCOPD PBMCCryopreservedSingle donor5×10⁷ cells (Vial)Human, adultLog in for availabilityEach$1,671.18Supplier Number: CPD-LKLT-50MItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product