Order Entry
Canada
Orders LinkContactUsLinkComponent
Acetone ACS

Acetone ACS

New Product
Supplier:   CHAPTEC INC CA
Acetone ACS
Supplier:  CHAPTEC INC CA
New Product
Certificates
Acetone ACS

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Acetone organic solvent A.C.S grade.

  • Acetone is colorless and mixes with water, alcohol, chloroform, ether and most oils
  • Evaporates quickly
  • Acetone is mainly used as a common solvent in organic reactions
  • For general and organic chemistry applications
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $77.65
    $77.65
    Each(1/Itm/Bx)
       $77.65

    Supplier Number: ABX7000-1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • Plastic bottle
    500 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $63.07
    $63.07
    Each(1/Itm/Bx)
       $63.07

    Supplier Number: ABX7000-500
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.