Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Dextran sulfate

Supplier:   G-Biosciences
Dextran sulfate
Supplier:  G-Biosciences
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Dextran sulfate sodium (CAS 9011‑18‑1) is a versatile high‑molecular‑weight polyanion, essential in molecular biology (e.g., hybridization), biochemistry (e.g., protein/lipoprotein precipitation), virology, inflammation modeling, and nanomedicine construction.

  • White to off-white crystalline powder BSE/TSE Free Loss on Drying: ≤10 % Free Sulfate: ≤0.5 % pH (1%, Water) at 25 °C: 5.0 to 7.5 Chloride: ≤1000 ppm Sulfur Content: 17.0 to 20.0 %
Synonyms:  Dextran sulfate sodium from Dextran, Dextran sulphate sodium from Dextran
  • CAS Number:9011-18-1
  • Molecular Weight:>500.000 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $773.35
    $773.35
    Each (50g)
       $773.35

    Supplier Number: RC-043
  • Bottle
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $3,288.20
    $3,288.20
    Each (250g)
       $3,288.20

    Supplier Number: RC-044

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.