Orders LinkContactUsLinkComponent
Multi-Use Press for 48 Filter Vials
Multi-Use Press for 48 Filter Vials
Multi-use press for 48 filter vials, metal, non sterile
Catalog # 76808-506
Supplier #:  35015
Multi-Use Press for 48 Filter Vials
Catalog # 76808-506
Multi-Use Press for 48 Filter Vials
Multi-Use Press for 48 Filter Vials
Multi-use press for 48 filter vials, metal, non sterile
Price
$2,290.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The Thomson multi-use press quickly depresses autosampler-ready filter vials.
  • Presses up to 48 autosampler ready filter vials at a time
  • Works with 48 position block; block fits some autosamplers
  • Easily automate filter vial pressing
  • Works with all Thomson filter vials
The 48 position block can be transferred to a robot for automation.
Specifications
  • Description:
    Multi-use press for 48 filter vials, metal, non sterile