Order Entry
Canada
Orders LinkContactUsLinkComponent
Phenol red 0,5% in aqueous solution

Phenol red 0,5% in aqueous solution

Phenol red 0,5% in aqueous solution
Supplier:  G-Biosciences
Certificates
Phenol red 0,5% in aqueous solution

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $98.60
    $98.60
    Each (100ml)
       $98.60

    Supplier Number: 786-562
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 250 ml
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $166.10
    $166.10
    Each (250ml)
       $166.10

    Supplier Number: 786-563
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.