Neuron Dissociation Solutions S
The product is ready-to-use without the need for preparations, and achieves simple isolation of neurons while maintaining high cell viability.
Supports high cell viability
- Simple and stable isolation of neurons
- Sterile and ready to Use
This is dispersion medium for primary neuronal cell cultures of rat and mouse and designed for the gentle and efficient isolation of neurons from neural tissue, supporting downstream cell culture applications.
Use this medium for dispersion of frozen nerve cells or fresh brain tissues dissected from rat and mouse.
This contains 10 sets of three different solutions of 2.5 ml each: enzyme solution, dispersion solution, and isolation solution.
- Media Format:Liquid
- Size:10 set
- Shelf life:18 months from date of manufacture
- Application:Dissociation of neural tissue for cell culture
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



