Order Entry
Canada
Orders LinkContactUsLinkComponent
 

pH Buffer Solutions

New Product
Supplier:   ACP Chemicals Inc
pH Buffer Solutions
Supplier:  ACP Chemicals Inc
New Product
Certificates
pH Buffer Solutions
pH Buffer Solutions

6 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Product Family Options

Product Information

  • pH accuracy
    pH buffer type
    Reference temperature
    Pack type
    Size
    pH
    Color
    Availability
    Price
  • CA11033-650
    Buffer pH 7, yellow poly bottle, 500 ml
    Buffer pH 7, yellow poly bottle, 500 ml
    ±0.01
    25 °C
    Plastic bottle
    500 ml
    7
    Yellow
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $62.68
    $62.68
    Each(1/Itm/Bx)
       $62.68

    Supplier Number: B5205-4L
    Specifications:
    New Product
    New Product
  • CA11033-624
    Buffer pH 10, blue poly bottle, 4 L
    Buffer pH 10, blue poly bottle, 4 L
    ±0.01
    Borate
    25 °C
    Plastic bottle
    4 L
    10
    Blue
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $507.51
    $507.51
    Each(1/Itm/Bx)
       $507.51

    Supplier Number: E1200-4L
    Specifications:
    New Product
    New Product
  • CA11033-592
    Buffer pH 10, blue poly bottle, 500 ml
    Buffer pH 10, blue poly bottle, 500 ml
    ±0.01
    Borate
    25 °C
    Plastic bottle
    500 ml
    10
    Blue
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $120.57
    $120.57
    Each(1/Itm/Bx)
       $120.57

    Supplier Number: H6100-2.5L
    Specifications:
    New Product
    New Product
  • CA11033-580
    Buffer pH 4, red poly bottle, 4 L
    Buffer pH 4, red poly bottle, 4 L
    ±0.01
    Potassium biphthalate 
    25 °C
    Plastic bottle
    4 L
    4
    Red
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $195.47
    $195.47
    Each(1/Itm/Bx)
       $195.47

    Supplier Number: P6211-20L
    Specifications:
    New Product
    New Product
  • CA11033-604
    Buffer pH 4, red poly bottle, 500 ml
    Buffer pH 4, red poly bottle, 500 ml
    ±0.01
    Potassium biphthalate 
    25 °C
    Plastic bottle
    500 ml
    4
    Red
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $161.78
    $161.78
    Each(1/Itm/Bx)
       $161.78

    Supplier Number: S5662-500G
    Specifications:
    New Product
    New Product
  • CA11033-644
    Buffer pH 7, yellow poly bottle, 4 L
    Buffer pH 7, yellow poly bottle, 4 L
    ±0.01
    25 °C
    Plastic bottle
    4 L
    7
    Yellow
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $223.60
    $223.60
    Each(1/Itm/Bx)
       $223.60

    Supplier Number: S8781-2.5L
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.