Phenol red pH-indicator
Product Details & Documents
pH indicator widely used in culture media to identify pH changes from neutral (red) to acidic (yellow) values. Typically a change in color from red to yellow indicates high rates of cellular death when used in tissue cultures. Observed to act as a weak estrogen in the culture of breast cancer cells. Induced oogenesis from cells scraped from the surface of adult ovaries.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:143-74-8
- Molecular Weight:354.37 g/mol
Specifications
Specification Test Results
- Constituents::100% Phenol red
Specifications
- Size:25 g
Specifications
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



