Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Crystal violet Cation-based violet dye

25 g
Catalog # ABCA_AB143095-25G
New Product
Supplier:   Abcam
 :  AB143095-25G
Crystal violet Cation-based violet dye
Catalog # ABCA_AB143095-25G
Supplier:  Abcam
New Product
CAS Number:  548-62-9
Crystal violet Cation-based violet dye

25 g

CAS Number:  548-62-9
Price
$154.00
Log in for availability
Each (25g)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cation-based violet dye. Interacts with aqueous KI - I₂ during Gram staining to produce a precipitate. Histological stain for Gram-positive bacteria and DNA. Shows antibacterial, antifungal, and anthelmintic effects in vivo. .

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Molecular Weight:408 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Crystal Violet

Specifications

  • Size:
    25 g

Specifications