Order Entry
Canada
Orders LinkContactUsLinkComponent
Caffeic acid ≥98% NF-kappaB inhibitor

Caffeic acid ≥98% NF-kappaB inhibitor

New Product
Supplier:   Abcam
Caffeic acid ≥98% NF-kappaB inhibitor
Supplier:  Abcam
New Product
Certificates

2 Options Available Below

Product Details & Documents

(IC₅₀ = 0.85 mM). Blood-brain barrier permeable.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Molecular Weight:180.16 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Caffeic acid
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 250 mg
    Log in for availability
    Log in for availability
    Each (250mg)
    Each (250mg) - $123.00
    $123.00
    Each (250mg)
       $123.00
    Minimum order quantity is 1

    Supplier Number: AB120862-250MG
    Specifications:
    New Product
    New Product
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $205.00
    $205.00
    Each (1g)
       $205.00
    Minimum order quantity is 1

    Supplier Number: AB120862-1G
    Specifications:
    New Product
    New Product