Order Entry
Canada
Orders LinkContactUsLinkComponent
Polysorbate 20

Polysorbate 20

10 tablet
Catalog # ABCA_AB285411-10TA
New Product
Supplier:   Abcam
Supplier #:  AB285411-10TABLET
Polysorbate 20
Catalog # ABCA_AB285411-10TA
Supplier:  Abcam
New Product
CAS Number:  9005-64-5
Polysorbate 20

10 tablet

CAS Number:  9005-64-5
Price
$339.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A non-ionic detergent useful for the isolation of membrane-bound proteins under non-denaturing conditions.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Molecular Weight:522.7 g/mol
Specifications

Specification Test Results

  • Constituents::
    0.81% Sodium chloride, 0.05% Sorbitan monolaurate, ethoxylated, 0.02% Potassium chloride

Specifications

  • Size:
    10 tablet

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.