Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Metyrapone ≥98% cytochrome P450 inhibitor

New Product
Supplier:   Abcam
Metyrapone ≥98% cytochrome P450 inhibitor
Supplier:  Abcam
New Product
Certificates

2 Options Available Below

Product Details & Documents

Selective cytochrome P450 inhibitor (IC₅₀ = 1 μM). Blocks cortisol synthesis in vitroand in vivo. Neuroprotective. Cell and blood-brain barrier permeable.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  2-Methyl-1,2-di(pyridin-3-yl)propan-1-one
  • CAS Number:54-36-4
  • Molecular Weight:226.27 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Metyrapone
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $553.00
    $553.00
    Each (1g)
       $553.00
     

    Supplier Number: AB144618-1G
    Specifications:
    New Product
    New Product
  • 200 mg
    Log in for availability
    Log in for availability
    Each (200mg)
    Each (200mg) - $164.00
    $164.00
    Each (200mg)
       $164.00
     

    Supplier Number: AB144618-200MG
    Specifications:
    New Product
    New Product