Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Aphidicolin ≥98% DNA polymerase alpha, delta and epsilon inhibitor

New Product
Supplier:   Abcam
Aphidicolin ≥98% DNA polymerase alpha, delta and epsilon inhibitor
Supplier:  Abcam
New Product
Certificates
Aphidicolin ≥98% DNA polymerase alpha, delta and epsilon inhibitor

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

DNA polymerase α, δ and ε inhibitor. Tetracyclic diterpene antibiotic. Reversible inhibitor of eukaryotic nuclear DNA replication. It blocks the cell cycle at early S phase.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (3R,4R,4aR,6aS,8R,9R,11aS,11bS)-4,9-Bis(hydroxymethyl)-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta[a]naphthalene-3,9-diol
  • CAS Number:38966-21-1
  • Molecular Weight:338.5 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    98% Aphidicolin from Nigrospora sphaerica
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $298.00
    $298.00
    Each (1mg)
       $298.00
     

    Supplier Number: AB142400-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • 5 mg
    Log in for availability
    Log in for availability
    Each (5mg)
    Each (5mg) - $1,120.00
    $1,120.00
    Each (5mg)
       $1,120.00
     

    Supplier Number: AB142400-5MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product