Orders LinkContactUsLinkComponent
LY-341495 ≥98% Group II antagonist
New Product
 :   Abcam
LY-341495 ≥98% Group II antagonist
 :  Abcam
New Product
LY-341495 ≥98% Group II antagonist
Water-soluble and very selective nanomolar potent antagonist for group II mGlu receptors. Also a relatively potent antagonist for group III mGlu receptors at high nanomolar to low micromolar concentrations. Disodium salt of LY 341495 which displays Kd values of 1.67 (human mGlu₂), 0.75 (mGlu₃), 31.6 (mGlu₆), 72.7 (mGlu₇a) and 14 (human mGlu₈a receptors) nM. LY 341495 displays IC₅₀ values of 7.8, 8.2 and 22 μM for mGlu₁a, mGlu₅a and mGlu₄ receptors, respectively. It is readily brain penetrant and active in vivo. Also available in simple stock solutions - add 1 ml of water to get an exact, ready-to-use concentration.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  (2S)-2-Amino-2-[(1S,2S)-2-carboxycycloprop-1-yl]-3-(xanth-9-yl) propanoic acid, 2-[(1s,2s)-2-Carboxycyclopropyl]-3-(9h-Xanthen-9-Yl)-D-Alanine, (1S,2S)-2-((S)-1-amino-1-carboxy-2-(9H-xanthen-9-yl)ethyl)cyclopropanecarboxylic acid
  •  :201943-63-7
  •  :353.4 g/mol
  • Constituents::
    100% LY 341495
Product Family Options
  • Size
  • 10 mg
    Each (10mg)
    Each (10mg) - $594.00
    $594.00
    Each (10mg)
       $594.00
     : AB120199-10MG
    New Product
    New Product
  • 50 mg
    Each (50mg)
    Each (50mg) - $2,470.00
    $2,470.00
    Each (50mg)
       $2,470.00
     : AB120199-50MG
    New Product
    New Product
  • 1 mg
    Each (1mg)
    Each (1mg) - $236.00
    $236.00
    Each (1mg)
       $236.00
     : AB120199-1MG