CDN Thymine-alpha,alpha,alpha,6-d4
New Product
Supplier: LGC Standards
About this item
2 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 78079-215
- 78079-216
- CAS No.:
- 200496-79-3
- 200496-79-3
- Formula:
- C₅D₄H₂N₂O₂
- C₅D₄H₂N₂O₂
- Molecular weight:
- 130.13800000000001
- 130.13800000000001
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 78079-215Thymine-alpha,alpha,alpha,6-d4Supplier Number: CDN-D-3362-0.5GSpecifications:New Product
More
Specifications
Cat. No.78079-215CAS No.200496-79-3FormulaC₅D₄H₂N₂O₂Molecular weight130.13800000000001New Product - 78079-216Thymine-alpha,alpha,alpha,6-d4Supplier Number: CDN-D-3362-1GSpecifications:New Product
More
Specifications
Cat. No.78079-216CAS No.200496-79-3FormulaC₅D₄H₂N₂O₂Molecular weight130.13800000000001New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



