Orders LinkContactUsLinkComponent
Multi-Function Laboratory Timer
Multi-Function Laboratory Timer
Multi-function timer, 6×9×7"
Catalog # 77973-346
New Product
Supplier #:  230123
Multi-Function Laboratory Timer
Catalog # 77973-346
New Product
Multi-Function Laboratory Timer
Multi-Function Laboratory Timer
Multi-function timer, 6×9×7"
Price
$100.07
Log in for availability
Case of 1
Product Details & Documents
Multi-function lab timers for accurate and efficient research to time multiple events simultaneously, track elapsed time, and manage time-sensitive procedures.
  • Multiple functions include timer, stopwatch, clock, and alarm
  • Easel stand, mounting magnet, and retractable hook for hanging
  • White finish
  • Uses one standard AAA battery (not included)
Improve the accuracy and reproducibility of experiments, especially when dealing with strict time schedules.
Specifications
  • Description:
    Multi-function timer
  • Dimensions:
    6×9×7"